Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vprbp
Entrez Gene ID
315987
Gene Name
Vpr (HIV-1) binding protein
Gene Aliases
RGD1305176
Chromosome Location
Chr.8: 115538365 - 115615282 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191798.1 NP_001178727.1 - 2077 62

View Full Product Details