Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc74a
Entrez Gene ID
680752
Gene Name
coiled-coil domain containing 74A
Gene Aliases
-
Chromosome Location
Chr.11: 87547027 - 87554513 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001058720.4 XP_001058720.2 4 - 5 2493 75

View Full Product Details