Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tp63
Entrez Gene ID
246334
Gene Name
tumor protein p63
Gene Aliases
Ket, P73l, Tp73l, Trp63
Chromosome Location
Chr.11: 78234853 - 78456329 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127339.1 NP_001120811.1 8 - 9 1274 61
NM_001127341.1 NP_001120813.1 8 - 9 1274 61
NM_001127342.1 NP_001120814.1 6 - 7 845 61
NM_001127343.1 NP_001120815.1 6 - 7 845 61
NM_001127344.1 NP_001120816.1 6 - 7 845 61
NM_019221.3 NP_062094.1 8 - 9 1274 61
XM_008768791.1 XP_008767013.1 9 - 10 1204 61
GenBank mRNA AJ277446.1 - 7 - 8 1076 61
AJ277447.1 - 6 - 7 845 61
AJ277448.1 - 7 - 8 1076 61
AJ277449.1 - 8 - 9 1127 61
AJ277450.1 - 6 - 7 845 61
AJ277451.1 - 8 - 9 1127 61
AJ277452.1 - 7 - 8 1076 61
AJ277453.1 - 6 - 7 845 61
Y10258.2 - 8 - 9 1274 61

View Full Product Details