Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tp63
Entrez Gene ID
246334
Gene Name
tumor protein p63
Gene Aliases
Ket, P73l, Tp73l, Trp63
Chromosome Location
Chr.11: 78234853 - 78456329 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127339.1 NP_001120811.1 10 - 11 1495 66
NM_001127342.1 NP_001120814.1 8 - 9 1066 66
NM_001127343.1 NP_001120815.1 8 - 9 1066 66
NM_019221.3 NP_062094.1 10 - 11 1495 66
XM_008768791.1 XP_008767013.1 11 - 12 1425 66
GenBank mRNA AJ277446.1 - 9 - 10 1297 66
AJ277447.1 - 8 - 9 1066 66
AJ277451.1 - 10 - 11 1348 66
AJ277452.1 - 9 - 10 1297 66
AJ277453.1 - 8 - 9 1066 66
Y10258.2 - 10 - 11 1495 66

View Full Product Details