Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spata24
Entrez Gene ID
291676
Gene Name
spermatogenesis associated 24
Gene Aliases
RGD1311742
Chromosome Location
Chr.18: 28446592 - 28454725 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025636.2 NP_001020807.2 3 - 4 324 91
XM_006254545.2 XP_006254607.1 3 - 4 443 91
GenBank mRNA BQ207964.1 - 1 - 2 91
DQ080016.1 - 3 - 4 324 91

View Full Product Details