Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spata24
Entrez Gene ID
291676
Gene Name
spermatogenesis associated 24
Gene Aliases
RGD1311742
Chromosome Location
Chr.18: 28446592 - 28454725 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006254545.2 XP_006254607.1 5 - 6 616 74

View Full Product Details