Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Giot1
Entrez Gene ID
171090
Gene Name
gonadotropin inducible ovarian transcription factor 1
Gene Aliases
-
Chromosome Location
Chr.7: 13104179 - 13122614 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133563.1 NP_598247.1 1 - 2 293 130
GenBank mRNA AB047636.1 - 1 - 2 293 130

View Full Product Details