Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gli2
Entrez Gene ID
304729
Gene Name
GLI family zinc finger 2
Gene Aliases
-
Chromosome Location
Chr.13: 34829021 - 34997009 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107169.1 NP_001100639.1 5 - 6 911 107
XM_008769455.1 XP_008767677.1 6 - 7 1532 107

View Full Product Details