Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nkd2
Entrez Gene ID
308068
Gene Name
naked cuticle homolog 2 (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.1: 32054412 - 32085416 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107454.1 NP_001100924.1 5 - 6 525 67
XM_008758691.1 XP_008756913.1 1 - 2 1223 67
XM_008758692.1 XP_008756914.1 8 - 9 2259 67
XM_008758693.1 XP_008756915.1 5 - 6 1040 67

View Full Product Details