Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gimap5
Entrez Gene ID
246774
Gene Name
GTPase, IMAP family member 5
Gene Aliases
IAN4, Ian4l1, Ian5, lyp
Chromosome Location
Chr.4: 78377228 - 78386672 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033913.1 NP_001029085.1 3 - 3 428 110
XM_006236457.2 XP_006236519.1 3 - 3 791 110
GenBank mRNA AY055777.2 - 1 - 1 140 110
AY550018.1 - 3 - 3 428 110
AY550021.1 - 5 - 5 946 110
BC092561.1 - 3 - 3 282 110

View Full Product Details