Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cuedc2
Entrez Gene ID
294009
Gene Name
CUE domain containing 2
Gene Aliases
-
Chromosome Location
Chr.1: 266078107 - 266087202 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079886.1 NP_001073355.1 6 - 7 1090 128
XM_006231458.2 XP_006231520.1 6 - 7 680 128
XM_006231459.2 XP_006231521.1 6 - 7 1143 128
XM_006231460.2 XP_006231522.1 6 - 7 686 128
XM_006231462.2 XP_006231524.2 6 - 7 1499 128
XM_006231465.2 XP_006231527.1 6 - 7 626 128
XM_006231466.2 XP_006231528.1 6 - 7 632 128
XM_008760408.1 XP_008758630.1 6 - 7 1496 128
XM_008760409.1 XP_008758631.1 6 - 7 1311 128
XM_008760410.1 XP_008758632.1 5 - 6 1414 128
XM_008760411.1 XP_008758633.1 6 - 7 1510 128
XM_008760412.1 XP_008758634.1 7 - 8 729 128
GenBank mRNA BC127518.1 - 6 - 7 1090 128
FQ220195.1 - 6 - 7 594 128
FQ229647.1 - 5 - 6 539 128

View Full Product Details