Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cpn1
Entrez Gene ID
365466
Gene Name
carboxypeptidase N, polypeptide 1
Gene Aliases
Cpn
Chromosome Location
Chr.1: 263733887 - 263762758 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053526.2 NP_445978.1 3 - 4 770 55
GenBank mRNA AB042599.1 - 3 - 4 779 55
BC088124.1 - 3 - 4 770 55
FQ219519.1 - 1 - 2 55 55

View Full Product Details