Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vsig8
Entrez Gene ID
289236
Gene Name
V-set and immunoglobulin domain containing 8
Gene Aliases
RGD1562464
Chromosome Location
Chr.13: 90943255 - 90951443 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105972.1 NP_001099442.1 3 - 4 576 99

View Full Product Details