Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smyd2
Entrez Gene ID
289372
Gene Name
SET and MYND domain containing 2
Gene Aliases
HSKM-B
Chromosome Location
Chr.13: 108642156 - 108686293 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_206851.1 NP_996733.1 5 - 6 570 90

View Full Product Details