The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191664.1 | NP_001178593.1 | 1 - 2 | 136 | 66 |
XM_006248867.1 | XP_006248929.1 | 3 - 4 | 435 | 66 | |
XM_006248868.1 | XP_006248930.1 | 3 - 4 | 432 | 66 |