Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mad1l1
Entrez Gene ID
680006
Gene Name
MAD1 mitotic arrest deficient-like 1 (yeast)
Gene Aliases
-
Chromosome Location
Chr.12: 16410083 - 16719824 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109387.1 NP_001102857.1 10 - 11 1127 65

View Full Product Details