The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106007.1 | NP_001099477.1 | 1 - 2 | 150 | 97 |
XM_006250996.2 | XP_006251058.1 | 1 - 2 | 158 | 97 | |
GenBank mRNA | BC160893.1 | - | 1 - 2 | 155 | 97 |