The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034198.2 | NP_001029370.1 | 7 - 8 | 774 | 149 |
NM_021671.3 | NP_067703.1 | 7 - 8 | 774 | 149 | |
GenBank mRNA | AB006135.1 | - | 7 - 8 | 732 | 149 |
BF552634.1 | - | 2 - 3 | 106 | 149 |