Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem33
Entrez Gene ID
59303
Gene Name
transmembrane protein 33
Gene Aliases
-
Chromosome Location
Chr.14: 42537352 - 42559580 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034198.2 NP_001029370.1 8 - 9 895 160
GenBank mRNA BF552634.1 - 3 - 4 227 160

View Full Product Details