Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Supt16h
Entrez Gene ID
305851
Gene Name
suppressor of Ty 16 homolog (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.15: 28574841 - 28611959 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107261.1 NP_001100731.1 17 - 18 2119 84

View Full Product Details