Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Apex1
Entrez Gene ID
79116
Gene Name
APEX nuclease (multifunctional DNA repair enzyme) 1
Gene Aliases
APE, Apex, REF-1
Chromosome Location
Chr.15: 27849943 - 27852083 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF311054.1 - 3 - 4 424 95

View Full Product Details