Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcfl5
Entrez Gene ID
311715
Gene Name
transcription factor-like 5 (basic helix-loop-helix)
Gene Aliases
RGD1305899
Chromosome Location
Chr.3: 176124846 - 176144531 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008762597.1 XP_008760819.1 4 - 5 1210 65

View Full Product Details