Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mknk1
Entrez Gene ID
500526
Gene Name
MAP kinase-interacting serine/threonine kinase 1
Gene Aliases
-
Chromosome Location
Chr.5: 134691870 - 134730887 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044267.1 NP_001037732.1 6 - 7 507 70
XM_006238694.2 XP_006238756.1 6 - 7 2053 70
XM_006238695.2 XP_006238757.1 6 - 7 2055 70
XM_006238696.2 XP_006238758.1 9 - 10 868 70
XM_006238697.2 XP_006238759.1 8 - 9 820 70
XM_006238698.2 XP_006238760.1 8 - 9 761 70
XM_006238699.2 XP_006238761.1 6 - 7 1987 70
GenBank mRNA BC098754.1 - 6 - 7 507 70

View Full Product Details