Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ipp
Entrez Gene ID
298439
Gene Name
intracisternal A particle-promoted polypeptide
Gene Aliases
-
Chromosome Location
Chr.5: 135351765 - 135390363 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106679.2 NP_001100149.2 5 - 6 1187 92
XM_006238640.2 XP_006238702.1 6 - 7 1359 92
XM_006238642.2 XP_006238704.1 6 - 7 1370 92
XM_006238643.2 XP_006238705.1 6 - 7 1371 92
XM_006238644.2 XP_006238706.1 6 - 7 1371 92
XM_008763992.1 XP_008762214.1 6 - 7 1370 92
GenBank mRNA FQ225549.1 - 4 - 5 489 92

View Full Product Details