Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dapk2
Entrez Gene ID
300799
Gene Name
death-associated protein kinase 2
Gene Aliases
-
Chromosome Location
Chr.8: 71822065 - 71941941 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006243306.2 XP_006243368.1 2 - 3 240 78
XM_008766241.1 XP_008764463.1 2 - 3 250 78

View Full Product Details