The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025123.3 | NP_001020294.3 | 5 - 6 | 1113 | 64 |
GenBank mRNA | BC093594.1 | - | 5 - 6 | 1113 | 64 |
BC100617.1 | - | 5 - 6 | 1112 | 64 |