Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Hspa8
- Entrez Gene ID
-
24468
- Gene Name
- heat shock 70kDa protein 8
- Gene Aliases
-
Hsc70
- Chromosome Location
-
Chr.8: 44989401 - 44993261 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_024351.2 |
NP_077327.1 |
1 - 2 |
78 |
72 |
GenBank mRNA |
BC061547.1 |
- |
1 - 2 |
85 |
72 |
- Gene Symbol
- LOC689955
- Entrez Gene ID
-
689955
- Gene Name
- similar to heat shock protein 8
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 34553275 - 34555388 on Build Rnor_6.0
View Full Product Details