Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zc3h12b
Entrez Gene ID
296864
Gene Name
zinc finger CCCH-type containing 12B
Gene Aliases
RGD1560891
Chromosome Location
Chr.X: 65040051 - 65074839 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002730248.4 XP_002730294.1 3 - 4 1043 86

View Full Product Details