Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ano8
Entrez Gene ID
306340
Gene Name
anoctamin 8
Gene Aliases
RGD1563946
Chromosome Location
Chr.16: 19884504 - 19894593 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006252965.2 XP_006253027.2 8 - 9 1167 132

View Full Product Details