Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pbx4
Entrez Gene ID
361131
Gene Name
pre-B-cell leukemia homeobox 4
Gene Aliases
Edg4
Chromosome Location
Chr.16: 21303117 - 21338771 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108399.1 NP_001101869.1 1 - 2 179 83

View Full Product Details