The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001044230.2 | NP_001037695.1 | 3 - 4 | 616 | 77 |
GenBank mRNA | DQ479969.1 | - | 3 - 4 | 594 | 77 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001044240.2 | NP_001037705.1 | 3 - 4 | 616 | 77 |