Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Map1s
Entrez Gene ID
290640
Gene Name
microtubule-associated protein 1S
Gene Aliases
Bpy2ip1, Mtap1s
Chromosome Location
Chr.16: 20268662 - 20278850 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008771085.1 XP_008769307.1 3 - 4 322 132
XM_008771086.1 XP_008769308.1 3 - 4 322 132

View Full Product Details