Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
St3gal5
Entrez Gene ID
83505
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 5
Gene Aliases
Siat9
Chromosome Location
Chr.4: 99937499 - 99999905 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031337.2 NP_112627.2 6 - 7 1336 80
XM_006236676.1 XP_006236738.1 5 - 6 997 80
XM_006236677.1 XP_006236739.1 6 - 7 1139 80
XM_006236679.2 XP_006236741.1 5 - 6 1130 80
XM_006236680.1 XP_006236742.1 5 - 6 997 80
XM_006236682.1 XP_006236744.1 4 - 5 824 80
XM_006236684.2 XP_006236746.1 4 - 5 1018 80
XM_008762996.1 XP_008761218.1 5 - 6 1159 80
XM_008762997.1 XP_008761219.1 4 - 5 1047 80
GenBank mRNA AB018049.1 - 5 - 6 1002 80
BC078798.1 - 6 - 7 1336 80

View Full Product Details