The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023972.3 | NP_076462.1 | 2 - 3 | 401 | 80 |
GenBank mRNA | AF240182.1 | - | 2 - 3 | 401 | 80 |
BC061548.1 | - | - | 413 | 80 | |
FQ213351.1 | - | - | 434 | 80 | |
FQ216190.1 | - | - | 434 | 80 | |
FQ216551.1 | - | - | 435 | 80 | |
FQ225681.1 | - | - | 434 | 80 | |
FQ228780.1 | - | - | 499 | 80 | |
FQ232358.1 | - | - | 434 | 80 |