Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chchd6
Entrez Gene ID
297436
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 6
Gene Aliases
-
Chromosome Location
Chr.4: 121340952 - 121565219 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106608.1 NP_001100078.1 7 - 8 880 86
GenBank mRNA FQ219980.1 - 7 - 8 861 86

View Full Product Details