Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pvrl1
Entrez Gene ID
192183
Gene Name
poliovirus receptor-related 1
Gene Aliases
HveC, nectin-1
Chromosome Location
Chr.8: 48094233 - 48157315 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006242963.2 XP_006243025.1 5 - 6 1810 114
XM_236210.8 XP_236210.3 5 - 6 1700 114

View Full Product Details