Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snx22
Entrez Gene ID
300796
Gene Name
sorting nexin 22
Gene Aliases
-
Chromosome Location
Chr.8: 71725405 - 71728730 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106832.1 NP_001100302.1 2 - 3 221 71
XM_006243304.2 XP_006243366.1 2 - 3 273 71
XM_006243305.2 XP_006243367.1 2 - 3 296 71
XM_008766240.1 XP_008764462.1 2 - 3 297 71

View Full Product Details