Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmprss5
Entrez Gene ID
266681
Gene Name
transmembrane protease, serine 5
Gene Aliases
Amp
Chromosome Location
Chr.8: 53411231 - 53430744 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153311.1 NP_695223.1 7 - 8 693 64
XM_008766194.1 XP_008764416.1 5 - 6 556 64
GenBank mRNA AF537098.1 - 7 - 8 693 64
AF537099.1 - 5 - 6 471 64

View Full Product Details