Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Serpinb13
Entrez Gene ID
304690
Gene Name
serpin peptidase inhibitor, clade B (ovalbumin), member 13
Gene Aliases
-
Chromosome Location
Chr.13: 27032048 - 27062620 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107168.1 NP_001100638.1 6 - 7 683 149
XM_008769454.1 XP_008767676.1 6 - 7 683 149

View Full Product Details