The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106537.1 | NP_001100007.1 | 2 - 3 | 530 | 78 |
XM_006235529.2 | XP_006235591.1 | 2 - 3 | 582 | 78 |