Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp24a1
Entrez Gene ID
25279
Gene Name
cytochrome P450, family 24, subfamily a, polypeptide 1
Gene Aliases
25-hydroxyvitaminD324-hydroxylase, Cyp24, VITD12
Chromosome Location
Chr.3: 168097484 - 168111917 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201635.2 NP_963966.1 10 - 11 1437 71
XM_006235672.2 XP_006235734.1 10 - 11 1779 71
GenBank mRNA BC100059.1 - 10 - 11 1782 71
X59506.1 - 10 - 11 1780 71

View Full Product Details