Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnaja3
Entrez Gene ID
360481
Gene Name
DnaJ (Hsp40) homolog, subfamily A, member 3
Gene Aliases
Tid-1
Chromosome Location
Chr.10: 11059701 - 11085182 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038595.1 NP_001033684.1 7 - 8 997 61
NM_001038596.1 NP_001033685.1 7 - 8 997 61
XM_006245817.2 XP_006245879.1 7 - 8 1037 61
GenBank mRNA AF516338.1 - 7 - 8 997 61
AY077460.1 - 7 - 8 997 61
AY264842.1 - 6 - 7 844 61

View Full Product Details