Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pam16
Entrez Gene ID
679907
Gene Name
presequence translocase-associated motor 16 homolog (S. cerevisiae)
Gene Aliases
Magmas
Chromosome Location
Chr.10: 11146359 - 11153936 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100136.2 NP_001093606.2 1 - 2 79 116
GenBank mRNA BG379323.1 - 4 - 5 116
FQ220566.1 - 1 - 2 100 116

View Full Product Details