The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100136.2 | NP_001093606.2 | 4 - 5 | 362 | 124 |
GenBank mRNA | AA933157.1 | - | 3 - 4 | 296 | 124 |
BE098288.1 | - | 3 - 4 | 296 | 124 | |
BG379323.1 | - | 1 - 2 | 124 | ||
FQ211057.1 | - | 4 - 5 | 317 | 124 | |
FQ220566.1 | - | 4 - 5 | 383 | 124 |