Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cluap1
Entrez Gene ID
363544
Gene Name
clusterin associated protein 1
Gene Aliases
-
Chromosome Location
Chr.10: 11847058 - 11884183 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014225.1 NP_001014247.1 8 - 9 1196 89
XM_006245832.2 XP_006245894.1 8 - 9 959 89
XM_008767512.1 XP_008765734.1 8 - 9 892 89
XM_008767513.1 XP_008765735.1 8 - 9 1254 89
XM_008767514.1 XP_008765736.1 7 - 8 796 89
XM_008767515.1 XP_008765737.1 7 - 8 800 89
XM_008767516.1 XP_008765738.1 7 - 8 798 89
XM_008767517.1 XP_008765739.1 6 - 7 661 89
XM_008767518.1 XP_008765740.1 6 - 7 718 89
GenBank mRNA BC079022.1 - 8 - 9 1196 89

View Full Product Details