The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014225.1 | NP_001014247.1 | 11 - 12 | 1432 | 138 |
XM_006245832.2 | XP_006245894.1 | 11 - 12 | 1195 | 138 | |
XM_008767512.1 | XP_008765734.1 | 11 - 12 | 1128 | 138 | |
XM_008767513.1 | XP_008765735.1 | 11 - 12 | 1490 | 138 | |
XM_008767514.1 | XP_008765736.1 | 10 - 11 | 1032 | 138 | |
XM_008767515.1 | XP_008765737.1 | 10 - 11 | 1036 | 138 | |
XM_008767516.1 | XP_008765738.1 | 10 - 11 | 1034 | 138 | |
XM_008767517.1 | XP_008765739.1 | 9 - 10 | 897 | 138 | |
XM_008767518.1 | XP_008765740.1 | 9 - 10 | 954 | 138 | |
GenBank mRNA | BC079022.1 | - | 11 - 12 | 1432 | 138 |