The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108634.1 | NP_001102104.1 | 2 - 3 | 363 | 75 |
GenBank mRNA | FQ213769.1 | - | 2 - 3 | 337 | 75 |