The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013431.1 | NP_001013449.1 | 2 - 3 | 293 | 76 |
GenBank mRNA | BC091407.1 | - | 2 - 3 | 293 | 76 |
FQ220478.1 | - | 2 - 3 | 158 | 76 | |
FQ220795.1 | - | 2 - 3 | 188 | 76 | |
FQ224864.1 | - | 2 - 3 | 206 | 76 |