Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wnt7a
Entrez Gene ID
114850
Gene Name
wingless-type MMTV integration site family, member 7A
Gene Aliases
-
Chromosome Location
Chr.4: 122994425 - 123040609 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100473.1 NP_001093943.1 1 - 2 327 62

View Full Product Details