Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rpusd3
Entrez Gene ID
362416
Gene Name
RNA pseudouridylate synthase domain containing 3
Gene Aliases
RGD1307582
Chromosome Location
Chr.4: 145362653 - 145370992 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108641.1 NP_001102111.1 3 - 4 553 66
XM_006237044.2 XP_006237106.1 4 - 5 418 66
XM_006237045.2 XP_006237107.1 4 - 5 593 66
XM_006237046.2 XP_006237108.1 4 - 5 418 66

View Full Product Details